Assay Details
Gene Symbol: TIMM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003353442.5 | XP_003353490.2 | 861 | 93 | ||
GenBank mRNA | AK234450.1 | - | 4 - 5 | 822 | 93 | |
AK391499.1 | - | 4 - 5 | 843 | 93 | ||
AY610071.1 | - | 4 - 5 | 825 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
