This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06875576_m1
See other ARL2 GE Assays ›
Gene Symbol:
ARL2
Gene Name:
ADP ribosylation factor like GTPase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 7157963 - 7167024 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06875576_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003122549.5 XP_003122597.4 266 86
XM_013994122.2 XP_013849576.1 335 86
GenBank mRNA AK234048.1 - 2 - 3 209 86
AK347570.1 - 2 - 3 177 86
AK392282.1 - 2 - 3 217 86
AY610137.1 - 2 - 3 200 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header