This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06875559_m1
See other NDUFS7 GE Assays ›
Gene Symbol:
NDUFS7
Gene Name:
NADH:ubiquinone oxidoreductase core subunit S7
Gene Aliases:
-
Chromosome Location:
Chr.2: 77080311 - 77087141 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06875559_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFS7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003353971.4 XP_003354019.2 195 72
XM_005661401.3 XP_005661458.2 195 72
XM_021084252.1 XP_020939911.1 317 72
XM_021084253.1 XP_020939912.1 317 72
GenBank mRNA AK234026.1 - 2 - 3 72 72
AK346668.1 - 2 - 3 77 72
AK397569.1 - 2 - 3 80 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_021084252.1, XM_021084253.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0