Assay Details
Gene Symbol: VSIG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924427.5 | XP_001924462.2 | 2235 | 70 | ||
XM_013982901.2 | XP_013838355.1 | 1428 | 70 | |||
XM_021073841.1 | XP_020929500.1 | 1708 | 70 | |||
XM_021073842.1 | XP_020929501.1 | 1689 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
