Assay Details
Gene Symbol: ABITRAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924196.7 | XP_001924231.1 | 446 | 77 |

Gene Symbol: ABITRAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001924196.7 | XP_001924231.1 | 446 | 77 |