This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06870211_g1
See other CERS5 GE Assays ›
Gene Symbol:
CERS5
Gene Name:
ceramide synthase 5
Gene Aliases:
-
Chromosome Location:
Chr.5: 16038918 - 16070103 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06870211_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERS5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021091642.1 XP_020947301.1 350 64
XM_021091643.1 XP_020947302.1 354 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0