This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06868181_m1
See other PGAM5 GE Assays ›
Gene Symbol:
PGAM5
Gene Name:
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Gene Aliases:
-
Chromosome Location:
Chr.14: 22773135 - 22780904 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06868181_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021073940.1 XP_020929599.1 630 59
XM_021073941.1 XP_020929600.1 630 59
GenBank mRNA AK233529.1 - 2 - 3 375 59
AK398114.1 - 2 - 3 631 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

isomerase mutase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0