This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06867750_g1
See other SLC6A12 GE Assays ›
Gene Symbol:
SLC6A12
Gene Name:
solute carrier family 6 member 12
Gene Aliases:
-
Chromosome Location:
Chr.5: 67550285 - 67573858 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06867750_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC6A12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003126574.5 XP_003126622.1 734 61
XM_005664091.3 XP_005664148.1 958 61
XM_005664092.3 XP_005664149.1 741 61
XM_013997850.2 XP_013853304.1 1067 61
XM_013997851.2 XP_013853305.1 843 61
XM_021092481.1 XP_020948140.1 1482 61
XM_021092483.1 XP_020948142.1 824 61
XM_021092484.1 XP_020948143.1 1067 61
GenBank mRNA AK233342.1 - 5 - 6 832 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0