This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06866705_m1
See other DHRS1 GE Assays ›
Gene Symbol:
DHRS1
Gene Name:
dehydrogenase/reductase 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 74992120 - 75002154 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06866705_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003128543.4 XP_003128591.1 1012 74
XM_005666234.3 XP_005666291.1 1009 74
GenBank mRNA AK232868.1 - 7 - 8 805 74
AK238438.1 - 6 - 7 822 74
AK394900.1 - 7 - 8 996 74
AK398307.1 - 7 - 8 772 74
AY609873.1 - 7 - 8 920 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0