Assay Details
Gene Symbol: GPR156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003132650.4 | XP_003132698.1 | 2555 | 60 | ||
XM_021070310.1 | XP_020925969.1 | 1455 | 60 | |||
XM_021070311.1 | XP_020925970.1 | 1636 | 60 | |||
XM_021070312.1 | XP_020925971.1 | 1791 | 60 | |||
XM_021070313.1 | XP_020925972.1 | 1549 | 60 | |||
XM_021070314.1 | XP_020925973.1 | 1530 | 60 | |||
XM_021070315.1 | XP_020925974.1 | 1302 | 60 | |||
XM_021070317.1 | XP_020925976.1 | 1367 | 60 | |||
XM_021070318.1 | XP_020925977.1 | 1624 | 60 | |||
XM_021070319.1 | XP_020925978.1 | 696 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
