This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06854774_m1
See other CEACAM18 GE Assays ›
Gene Symbol:
CEACAM18
Gene Name:
CEA cell adhesion molecule 18
Gene Aliases:
-
Chromosome Location:
Chr.6: 55749224 - 55759788 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06854774_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021094855.1 XP_020950514.1 800 68
XM_021094856.1 XP_020950515.1 792 68
XM_021094857.1 XP_020950516.1 774 68
XM_021094858.1 XP_020950517.1 790 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Sus scrofa

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0