This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06854688_g1
See other CHST8 GE Assays ›
Gene Symbol:
CHST8
Gene Name:
carbohydrate sulfotransferase 8
Gene Aliases:
-
Chromosome Location:
Chr.6: 43371436 - 43493250 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06854688_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHST8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021094298.1 XP_020949957.1 1152 72
XM_021094299.1 XP_020949958.1 686 72
XM_021094300.1 XP_020949959.1 1009 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header