Assay Details
Gene Symbol: CABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021072790.1 | XP_020928449.1 | 567 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: CABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021072790.1 | XP_020928449.1 | 567 | 52 |
Set Membership: |
![]() ![]() ![]() ![]() |