Assay Details
Gene Symbol: IER5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674588.3 | XP_005674645.2 | 1219 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: IER5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674588.3 | XP_005674645.2 | 1219 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() |