Assay Details
Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005665993.3 | XP_005666050.1 | 1466 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005665993.3 | XP_005666050.1 | 1466 | 111 |
Set Membership: |
![]() ![]() ![]() |