This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06825186_m1
See other THPO GE Assays ›
Gene Symbol:
THPO
Gene Name:
thrombopoietin
Gene Aliases:
MGDF, ML
Chromosome Location:
Chr.13: 122259654 - 122267860 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06825186_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: THPO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003483282.4 XP_003483330.2 1283 69
XM_005670050.3 XP_005670107.1 1726 69
XM_021069969.1 XP_020925628.1 1283 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
thrombopoietin-mediated signaling pathway
positive regulation of megakaryocyte differentiation
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
cytokine activity
hormone activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0