This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04954747_m1
See other NKX2-1 GE Assays ›
Gene Symbol:
NKX2-1
Gene Name:
NK2 homeobox 1
Gene Aliases:
TTF1
Chromosome Location:
Chr.7: 63487200 - 63491580 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04954747_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NKX2-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001311186.1 NP_001298115.1 2 - 3 561 96
NM_001311187.1 NP_001298116.1 1 - 2 714 96
XM_005666203.3 XP_005666260.1 655 96
XM_021098030.1 XP_020953689.1 699 96
GenBank mRNA EW050982.2 - 1 - 2 151 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0