Assay Details
Gene Symbol: NR1H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287412.1 | NP_001274341.1 | 8 - 9 | 1248 | 68 | |
XM_001928800.7 | XP_001928835.4 | 1496 | 68 | |||
XM_005664208.3 | XP_005664265.2 | 1255 | 68 | |||
XM_005664209.3 | XP_005664266.2 | 1483 | 68 | |||
XM_005664210.3 | XP_005664267.1 | 1168 | 68 | |||
XM_013998033.2 | XP_013853487.1 | 1156 | 68 | |||
XM_021090932.1 | XP_020946591.1 | 1485 | 68 | |||
XM_021090933.1 | XP_020946592.1 | 1470 | 68 | |||
XM_021090934.1 | XP_020946593.1 | 1390 | 68 | |||
XM_021090935.1 | XP_020946594.1 | 1234 | 68 | |||
XM_021090936.1 | XP_020946595.1 | 1222 | 68 | |||
XM_021090938.1 | XP_020946597.1 | 1467 | 68 | |||
XM_021090939.1 | XP_020946598.1 | 1399 | 68 | |||
GenBank mRNA | KF534517.1 | - | 8 - 9 | 1251 | 68 | |
KF597010.1 | - | 8 - 9 | 1209 | 68 | ||
KF597011.1 | - | 8 - 9 | 1221 | 68 | ||
KF597012.1 | - | 8 - 9 | 1176 | 68 | ||
KF597013.1 | - | 8 - 9 | 1188 | 68 | ||
KF597014.1 | - | 7 - 8 | 1110 | 68 | ||
KF597015.1 | - | 7 - 8 | 1122 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
