Assay Details
Gene Symbol: MIR9842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128529.1 | 1 | 38 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | colony stimulating factor 3 receptor | CSF3R | -382.9kb | Sense | Chr. 6 - 92550159 - 92565798 on NCBI Build Sscrofa11.1 | |
Downstream | zinc finger CCCH-type containing 12A | ZC3H12A | MCPIP1 | 592.6kb | Anti-sense | Chr. 6 - 93541334 - 93551200 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
