This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04652473_s1
See other MIR181A-2 GE Assays ›
Gene Symbol:
MIR181A-2
Gene Name:
microRNA mir-181a-2
Gene Aliases:
ssc-mir-181a-2
Chromosome Location:
Chr.1: 265497249 - 265497328 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04652473_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR181A-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_035385.1 1 47 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear receptor subfamily 6 group A member 1 NR6A1 GCNF Anti-sense Chr. 1 - 265320597 - 265570941 on NCBI Build Sscrofa11.1
Downstream microRNA mir-181b-2 MIR181B-2 MIR181B,
MIRN181B,
ssc-mir-181b,
ssc-mir-181b-2
1.8kb Sense Chr. 1 - 265499111 - 265499196 on NCBI Build Sscrofa11.1

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0