This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04647690_s1
See other MIR216-1 GE Assays ›
Gene Symbol:
MIR216-1
Gene Name:
microRNA mir-216-1
Gene Aliases:
MIR216, ssc-mir-216, ssc-mir-216-1
Chromosome Location:
Chr.3: 85334523 - 85334627 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04647690_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR216-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038486.1 1 46 79

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coiled-coil domain containing 85A CCDC85A Anti-sense Chr. 3 - 84958384 - 85649854 on NCBI Build Sscrofa11.1
Downstream microRNA mir-217-1 MIR217-1 MIR217,
ssc-mir-217,
ssc-mir-217-1
6.6kb Sense Chr. 3 - 85341255 - 85341361 on NCBI Build Sscrofa11.1

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0