This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04327011_m1
See other B4GALNT2 GE Assays ›
Gene Symbol:
B4GALNT2
Gene Name:
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 25368234 - 25410021 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04327011_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B4GALNT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244330.1 NP_001231259.1 4 - 5 496 62
XM_021066146.1 XP_020921805.1 587 62
XM_021066148.1 XP_020921807.1 587 62
XM_021066149.1 XP_020921808.1 587 62
XM_021066150.1 XP_020921809.1 587 62
XM_021066151.1 XP_020921810.1 587 62
GenBank mRNA AK236764.1 - 4 - 5 496 62
AK396108.1 - 4 - 5 592 62
KF501048.1 - 4 - 5 498 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header