This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04326628_m1
See other ARHGDIB GE Assays ›
Gene Symbol:
ARHGDIB
Gene Name:
Rho GDP dissociation inhibitor beta
Gene Aliases:
-
Chromosome Location:
Chr.5: 57576754 - 57598395 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04326628_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGDIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244240.1 NP_001231169.1 5 - 6 487 77
GenBank mRNA AK236902.1 - 5 - 6 471 77
AK237374.1 - 5 - 6 503 77
AK345764.1 - 4 - 5 280 77
AK351946.1 - 4 - 5 400 77
AK396003.1 - 5 - 6 470 77
AY610457.1 - 4 - 5 416 77
FD635764.1 - 5 - 6 491 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm