This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04326370_m1
See other SORD GE Assays ›
Gene Symbol:
SORD
Gene Name:
sorbitol dehydrogenase
Gene Aliases:
SDH, XDH
Chromosome Location:
Chr.1: 126659198 - 126701084 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04326370_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SORD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244162.1 NP_001231091.1 5 - 6 699 66
XM_021062686.1 XP_020918345.1 561 66
GenBank mRNA AK236025.1 - 5 - 6 659 66
AK236181.1 - 4 - 5 614 66
AK351174.1 - 4 - 5 581 66
AK389436.1 - 5 - 6 699 66
AK392246.1 - 5 - 6 687 66
AY610034.1 - 5 - 6 614 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

sperm motility
oxidation-reduction process
zinc ion binding
D-xylulose reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0