This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04326300_m1
See other MDH2 GE Assays ›
Gene Symbol:
MDH2
Gene Name:
malate dehydrogenase 2
Gene Aliases:
MDH, MMDH
Chromosome Location:
Chr.3: 10120121 - 10132705 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04326300_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244153.1 NP_001231082.1 6 - 7 684 62
GenBank mRNA AK232730.1 - 6 - 7 686 62
AK235773.1 - 6 - 7 752 62
AK235801.1 - 6 - 7 752 62
AK238803.1 - 6 - 7 749 62
AK394868.1 - 6 - 7 685 62
AK398042.1 - 6 - 7 743 62
AK398705.1 - 6 - 7 763 62
AK399962.1 - 6 - 7 763 62
AK400328.1 - 6 - 7 698 62
DQ402968.1 - 5 - 6 553 62
M16427.1 - 5 - 6 513 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
malate metabolic process
aerobic respiration
L-malate dehydrogenase activity
protein homodimerization activity
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0