This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04326039_g1
See other RPS11 GE Assays ›
Gene Symbol:
RPS11
Gene Name:
ribosomal protein S11
Gene Aliases:
-
Chromosome Location:
Chr.6: 54577414 - 54579797 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04326039_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244070.1 NP_001230999.1 3 - 4 499 158
XM_005664714.3 XP_005664771.2 388 158
GenBank mRNA AK233375.1 - 3 - 4 246 158
AK236665.1 - 804 158
AK347721.1 - 3 - 4 247 158
AK392151.1 - 3 - 4 304 158
AK392193.1 - 3 - 4 247 158
CJ013638.1 - 3 - 4 499 158

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0