This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325705_m1
See other MSR1 GE Assays ›
Gene Symbol:
MSR1
Gene Name:
macrophage scavenger receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 3859903 - 3939612 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325705_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243874.1 NP_001230803.1 2 - 3 157 63
XM_005672637.3 XP_005672694.2 411 63
XM_013984974.2 XP_013840428.2 508 63
XM_013984976.2 XP_013840430.2 314 63
XM_013984977.2 XP_013840431.2 313 63
XM_021077262.1 XP_020932921.1 603 63
XM_021077263.1 XP_020932922.1 506 63
XM_021077264.1 XP_020932923.1 409 63
GenBank mRNA AK230664.1 - 2 - 3 154 63
AK230809.1 - 2 - 3 154 63
AK391299.1 - 2 - 3 158 63
DB793168.1 - 2 - 3 158 63
EW333758.2 - 2 - 3 129 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0