This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04325586_g1
See other CRACR2B GE Assays ›
Gene Symbol:
CRACR2B
Gene Name:
calcium release activated channel regulator 2B
Gene Aliases:
EFCAB4A
Chromosome Location:
Chr.2: 518102 - 524085 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04325586_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRACR2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243838.1 NP_001230767.1 6 - 7 778 93
XM_013989677.2 XP_013845131.1 1456 93
XM_013989679.2 XP_013845133.1 1420 93
XM_013989682.2 XP_013845136.1 1456 93
XM_013989684.2 XP_013845138.1 1548 93
GenBank mRNA AK350005.1 - 6 - 7 778 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0