This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04324584_m1
See other CLDND1 GE Assays ›
Gene Symbol:
CLDND1
Gene Name:
claudin domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 160397444 - 160405675 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04324584_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLDND1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243498.1 NP_001230427.1 3 - 4 461 97
XM_005670258.3 XP_005670315.2 546 97
XM_005670259.3 XP_005670316.2 515 97
XM_005670260.3 XP_005670317.2 556 97
XM_005670262.3 XP_005670319.1 541 97
GenBank mRNA AK343803.1 - 3 - 4 461 97
AK392457.1 - 3 - 4 456 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0