This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04324187_g1
See other HDDC3 GE Assays ›
Gene Symbol:
HDDC3
Gene Name:
HD domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.7: 53525984 - 53527913 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04324187_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDDC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243409.1 NP_001230338.1 2 - 3 184 62
XM_021098000.1 XP_020953659.1 207 62
GenBank mRNA AK343282.1 - 2 - 3 324 62
AK392506.1 - 1 - 2 486 62
AK394867.1 - 2 - 3 286 62
AK395225.1 - 1 - 2 461 62
AK395620.1 - 2 - 3 184 62
FS689498.1 - 2 - 3 184 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0