This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04322085_m1
See other GRPEL2 GE Assays ›
Gene Symbol:
GRPEL2
Gene Name:
GrpE like 2, mitochondrial
Gene Aliases:
-
Chromosome Location:
Chr.2: 150510520 - 150520013 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04322085_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRPEL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244870.1 NP_001231799.1 3 - 4 319 70
XM_021078521.1 XP_020934180.1 342 70
GenBank mRNA CF176583.1 - 3 - 4 319 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0