This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04321698_m1
See other GZMA GE Assays ›
Gene Symbol:
GZMA
Gene Name:
granzyme A (granzyme 1, cytotoxic T-lymphocyte-associated serine esterase 3)
Gene Aliases:
-
Chromosome Location:
Chr.16: 34282160 - 34290422 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04321698_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GZMA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198926.1 NP_001185855.1 2 - 3 223 75
GenBank mRNA AK346600.1 - 2 - 3 247 75
HQ225831.1 - 2 - 3 223 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0