This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04246926_m1
See other ATP5F1A GE Assays ›
Gene Symbol:
ATP5F1A
Gene Name:
ATP synthase F1 subunit alpha
Gene Aliases:
ATP5A1, ATP5A2
Chromosome Location:
Chr.1: 95738789 - 95750835 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04246926_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5F1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001185142.1 NP_001172071.1 2 - 3 187 85
GenBank mRNA AK232528.1 - 2 - 3 192 85
AK236714.1 - 2 - 3 204 85
AK239102.1 - 2 - 3 204 85
AK396284.1 - 2 - 3 204 85
AK396428.1 - 2 - 3 202 85
AK396740.1 - 2 - 3 192 85
AK396809.1 - 2 - 3 227 85
AK397056.1 - 2 - 3 192 85
AY609645.1 - 2 - 3 187 85
EU650784.1 - 2 - 3 187 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0