This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss04245689_m1
See other CDK7 GE Assays ›
Gene Symbol:
CDK7
Gene Name:
cyclin dependent kinase 7
Gene Aliases:
-
Chromosome Location:
Chr.16: 47410136 - 47445795 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss04245689_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDK7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166314.1 NP_001159786.1 7 - 8 519 96
XM_021076536.1 XP_020932195.1 651 96
XM_021076537.1 XP_020932196.1 498 96
XM_021076538.1 XP_020932197.1 498 96
GenBank mRNA AB499891.1 - 7 - 8 519 96
AK239686.1 - 7 - 8 631 96
AK347057.1 - 7 - 8 642 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0