This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03822121_s1
See other APOA5 GE Assays ›
Gene Symbol:
APOA5
Gene Name:
apolipoprotein A5
Gene Aliases:
-
Chromosome Location:
Chr.9: 44177834 - 44180880 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03822121_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOA5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159308.1 NP_001152780.1 3 252 111
XM_005667368.3 XP_005667425.1 300 111
XM_005667369.3 XP_005667426.1 290 111
GenBank mRNA AK232735.1 - 3 260 111
AK345820.1 - 3 264 111
AK346104.1 - 4 392 111
AK394310.1 - 3 285 111
FJ810861.1 - 3 252 111
HM991258.1 - 3 265 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

apolipoprotein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0