This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03394253_m1
See other CD59 GE Assays ›
Gene Symbol:
CD59
Gene Name:
CD59 molecule
Gene Aliases:
-
Chromosome Location:
Chr.2: 27186587 - 27207303 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03394253_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214170.1 NP_999335.1 3 - 4 262 94
XM_005661031.3 XP_005661088.1 410 94
GenBank mRNA AF020302.1 - 3 - 4 262 94
AF058328.1 - 1 - 2 98 94
AK238416.1 - 3 - 4 226 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons

Gene Ontology Categories:

Function(s)

innate immune response

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0