This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03393473_m1
See other IL12B GE Assays ›
Gene Symbol:
IL12B
Gene Name:
interleukin 12B
Gene Aliases:
-
Chromosome Location:
Chr.16: 64143268 - 64158929 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03393473_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL12B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214013.1 NP_999178.1 6 - 7 895 68
GenBank mRNA U08317.1 - 6 - 7 895 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
positive regulation of T-helper 1 type immune response
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
negative regulation of inflammatory response to antigenic stimulus
cell cycle arrest
cell proliferation
response to UV-B
positive regulation of activation of Janus kinase activity
cell migration
natural killer cell activation
negative regulation of interleukin-10 production
negative regulation of interleukin-17 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell proliferation
positive regulation of smooth muscle cell apoptotic process
T-helper 1 cell cytokine production
T-helper cell differentiation
positive regulation of activated T cell proliferation
positive regulation of tyrosine phosphorylation of STAT protein
defense response to protozoan
positive regulation of osteoclast differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of protein secretion
defense response to Gram-negative bacterium
positive regulation of NK T cell proliferation
defense response to virus
cellular response to lipopolysaccharide
cellular response to interferon-gamma
negative regulation of vascular endothelial growth factor signaling pathway
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
cytokine activity
growth factor activity
identical protein binding
interleukin-23 receptor binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0