This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03393382_u1
See other IGFBP2 GE Assays ›
Gene Symbol:
IGFBP2
Gene Name:
insulin like growth factor binding protein 2
Gene Aliases:
pIGFBP-2
Chromosome Location:
Chr.15: 118822092 - 118850985 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03393382_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214003.1 NP_999168.1 4 951 71
GenBank mRNA AF120326.1 - 4 951 71
AK347326.1 - 4 969 71
AK395649.1 - 4 996 71
HQ432890.1 - 4 889 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

response to insulin
positive regulation of activated T cell proliferation
regulation of insulin-like growth factor receptor signaling pathway
positive regulation of growth
insulin-like growth factor I binding
insulin-like growth factor II binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0