This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03392461_u1
See other CD46 GE Assays ›
Gene Symbol:
CD46
Gene Name:
CD46 molecule, complement regulatory protein
Gene Aliases:
MCP
Chromosome Location:
Chr.9: 135025439 - 135078910 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03392461_u1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD46
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213888.1 NP_999053.1 9 1052 68
XM_021102197.1 XP_020957856.1 1122 68
XM_021102198.1 XP_020957857.1 1121 68
XM_021102199.1 XP_020957858.1 1121 68
GenBank mRNA AK344510.1 - 9 1099 68
D70897.1 - 9 1052 68
KJ513478.1 - 9 994 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Inventoried Amplicon spans exons and probe does not span exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
complement activation, classical pathway
single fertilization
innate immune response
negative regulation of complement activation, alternative pathway
negative regulation of complement activation, classical pathway
endopeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0