This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03392406_m1
See other SOX9 GE Assays ›
Gene Symbol:
SOX9
Gene Name:
SRY-box transcription factor 9
Gene Aliases:
SOX-9
Chromosome Location:
Chr.12: 8642154 - 8647315 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03392406_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOX9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213843.2 NP_999008.2 2 - 3 686 145
GenBank mRNA AF029696.1 - 2 - 3 686 145

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
cartilage condensation
cell fate specification
epithelial to mesenchymal transition
tissue homeostasis
positive regulation of protein phosphorylation
hair follicle development
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
negative regulation of immune system process
heart valve development
heart valve morphogenesis
endocardial cushion morphogenesis
chondrocyte differentiation involved in endochondral bone morphogenesis
chondrocyte hypertrophy
growth plate cartilage chondrocyte growth
nucleosome assembly
chromatin remodeling
signal transduction
epidermal growth factor receptor signaling pathway
spermatogenesis
positive regulation of cell proliferation
male gonad development
regulation of cell cycle process
positive regulation of gene expression
positive regulation of epithelial cell migration
neural crest cell fate specification
positive regulation of phosphatidylinositol 3-kinase signaling
male germ-line sex determination
cAMP-mediated signaling
negative regulation of ossification
positive regulation of epithelial cell differentiation
otic vesicle formation
negative regulation of chondrocyte differentiation
positive regulation of chondrocyte differentiation
somatic stem cell population maintenance
regulation of cell proliferation
regulation of apoptotic process
negative regulation of apoptotic process
protein kinase B signaling
negative regulation of myoblast differentiation
negative regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of fatty acid oxidation
negative regulation of photoreceptor cell differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
cartilage development
Sertoli cell differentiation
retina development in camera-type eye
retinal rod cell differentiation
epithelial cell proliferation involved in prostatic bud elongation
intestinal epithelial structure maintenance
regulation of cell proliferation involved in tissue homeostasis
regulation of cartilage development
positive regulation of cartilage development
morphogenesis of a branching epithelium
protein-containing complex assembly
negative regulation of biomineral tissue development
ERK1 and ERK2 cascade
response to fatty acid
cellular response to mechanical stimulus
cellular response to epidermal growth factor stimulus
cellular response to heparin
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
renal vesicle induction
metanephric nephron tubule formation
negative regulation of canonical Wnt signaling pathway
cochlea morphogenesis
positive regulation of kidney development
positive regulation of branching involved in ureteric bud morphogenesis
positive regulation of male gonad development
positive regulation of mesenchymal stem cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0