This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03391924_m1
See other SLC35A3 GE Assays ›
Gene Symbol:
SLC35A3
Gene Name:
solute carrier family 35 member A3
Gene Aliases:
S35A3
Chromosome Location:
Chr.4: 118102800 - 118153144 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03391924_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC35A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024588.1 NP_001019759.1 3 - 4 461 82
XM_005663640.3 XP_005663697.2 945 82
XM_005663641.3 XP_005663698.1 661 82
XM_021088749.1 XP_020944408.1 3776 82
GenBank mRNA AY996816.1 - 3 - 4 461 82
DQ883629.1 - 3 - 4 461 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0