This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03391812_m1
See other GBA GE Assays ›
Gene Symbol:
GBA
Gene Name:
glucosylceramidase beta
Gene Aliases:
-
Chromosome Location:
Chr.4: 94583905 - 94606689 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03391812_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GBA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005730.1 NP_001005730.1 9 - 10 1417 59
XM_013997000.2 XP_013852454.1 1785 59
GenBank mRNA AK233064.1 - 10 - 11 1723 59
AK233163.1 - 10 - 11 1723 59
AK234517.1 - 9 - 10 1521 59
AK397675.1 - 10 - 11 1759 59
AK401022.1 - 7 - 8 1194 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glucosylceramide catabolic process
autophagy
lysosome organization
cholesterol metabolic process
lipid glycosylation
regulation of TOR signaling
glucosylceramidase activity
glucosyltransferase activity
steryl-beta-glucosidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0