This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03390307_g1
See other CDK4 GE Assays ›
Gene Symbol:
CDK4
Gene Name:
cyclin dependent kinase 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 23039206 - 23041959 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03390307_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDK4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001123097.1 NP_001116569.1 7 - 8 907 73
GenBank mRNA AK239738.1 - 7 - 8 1056 73
AK390268.1 - 7 - 8 1093 73
AK395098.1 - 6 - 7 981 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
signal transduction
regulation of gene expression
positive regulation of G2/M transition of mitotic cell cycle
cellular response to insulin stimulus
regulation of multicellular organism growth
response to drug
regulation of insulin receptor signaling pathway
regulation of lipid biosynthetic process
positive regulation of fibroblast proliferation
regulation of lipid catabolic process
cell division
adipose tissue development
negative regulation of cell cycle arrest
cellular response to lipopolysaccharide
cellular response to interleukin-4
cellular response to phorbol 13-acetate 12-myristate
cellular response to ionomycin
ATP binding
cyclin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0