This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03390117_m1
See other MFGE8 GE Assays ›
Gene Symbol:
MFGE8
Gene Name:
milk fat globule EGF and factor V/VIII domain containing
Gene Aliases:
-
Chromosome Location:
Chr.7: 54539970 - 54554885 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03390117_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFGE8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122984.1 NP_001116456.1 7 - 8 1056 74
XM_013996945.2 XP_013852399.1 1143 74
XM_021098306.1 XP_020953965.1 1332 74
GenBank mRNA AK233926.1 - 8 - 9 1062 74
AK390171.1 - 7 - 8 1064 74
EU559724.1 - 7 - 8 1056 74
Y11683.1 - 942 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s)

angiogenesis
cell adhesion
single fertilization

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0