This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03389945_m1
See other SLA-DRA GE Assays ›
Gene Symbol:
SLA-DRA
Gene Name:
MHC class II DR-alpha
Gene Aliases:
SLA-DR-alpha, SLA-DRA1, SLA-DRA2
Chromosome Location:
Chr.7: 24825037 - 24830650 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03389945_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLA-DRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113706.1 NP_001107178.1 4 - 5 846 89
GenBank mRNA AK231578.1 - 4 - 5 991 89
AK239665.1 - 4 - 5 859 89
AK351169.1 - 4 - 5 848 89
AK351896.1 - 4 - 5 850 89
AK389739.1 - 4 - 5 869 89
AK394576.1 - 4 - 5 858 89
AK397711.1 - 4 - 5 847 89
AK398000.1 - 4 - 5 851 89
EF143987.1 - 4 - 5 785 89
EU728595.1 - 4 - 5 785 89
M93028.1 - 4 - 5 846 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0