This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03389890_m1
See other SLA-DQB1 GE Assays ›
Gene Symbol:
SLA-DQB1
Gene Name:
SLA-DQ beta1 domain
Gene Aliases:
SLA-DQB, SLADQB
Chromosome Location:
Chr.7: 24969034 - 24977238 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03389890_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLA-DQB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113694.1 NP_001107166.1 1 - 2 165 147
GenBank mRNA AB009659.1 - 1 - 2 147
AB016738.1 - 1 - 2 114 147
AB016741.1 - 1 - 2 147
AB016742.1 - 1 - 2 114 147
AB215114.1 - 1 - 2 147
AB215117.1 - 1 - 2 147
AF464024.1 - 1 - 2 147
AF464025.1 - 1 - 2 165 147
AF464028.1 - 1 - 2 147
AF464029.1 - 1 - 2 147
AK232216.1 - 1 - 2 147
AK239698.1 - 1 - 2 147
AK344947.1 - 1 - 2 147
AK346563.1 - 1 - 2 147
AK346688.1 - 1 - 2 147
AK346818.1 - 1 - 2 147
AK347729.1 - 1 - 2 147
AK350143.1 - 1 - 2 147
AK351451.1 - 1 - 2 147
AK351783.1 - 1 - 2 147
AK351935.1 - 1 - 2 147
AK392873.1 - 1 - 2 147
AK393066.1 - 1 - 2 147
AK398442.1 - 1 - 2 147
AK398920.1 - 1 - 2 147
AY102477.1 - 1 - 2 114 147
AY102478.1 - 1 - 2 147
AY135569.1 - 1 - 2 147
AY135570.1 - 1 - 2 147
AY243105.1 - 1 - 2 147
AY459300.1 - 1 - 2 147
DQ883219.1 - 1 - 2 147
EU170461.1 - 1 - 2 147
EU432063.1 - 1 - 2 147
EU432064.1 - 1 - 2 147
EU722914.1 - 1 - 2 147
EU722915.1 - 1 - 2 147 147
FJ905828.1 - 1 - 2 147
JQ511975.1 - 1 - 2 114 147
KC511016.1 - 1 - 2 179 147
KJ776454.1 - 1 - 2 147
KT194215.1 - 1 - 2 147
KT194216.1 - 1 - 2 147
KU754584.1 - 1 - 2 147
KU754587.1 - 1 - 2 147
M31497.1 - 1 - 2 165 147
M32117.1 - 1 - 2 165 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s)

adaptive immune response
antigen processing and presentation of peptide or polysaccharide antigen via MHC class II

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0