This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03388906_g1
See other COX7C GE Assays ›
Gene Symbol:
COX7C
Gene Name:
cytochrome c oxidase subunit 7C
Gene Aliases:
COX7C1
Chromosome Location:
Chr.2: 94345436 - 94347709 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03388906_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COX7C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097474.1 NP_001090943.1 2 - 3 308 102
GenBank mRNA AK231029.1 - 2 - 3 305 102
AK343779.1 - 2 - 3 305 102
AK392551.1 - 2 - 3 305 102
DQ456972.1 - 2 - 3 308 102
DQ456973.1 - 2 - 3 308 102
DQ456974.1 - 1 - 2 75 102
DQ629155.1 - 2 - 3 248 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

oxidase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

hydrogen ion transmembrane transport
cytochrome-c oxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0