This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03388488_m1
See other ALDH2 GE Assays ›
Gene Symbol:
ALDH2
Gene Name:
aldehyde dehydrogenase 2 family member
Gene Aliases:
-
Chromosome Location:
Chr.14: 39902317 - 39933113 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03388488_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044611.2 NP_001038076.1 3 - 4 452 58
GenBank mRNA AK232401.1 - 3 - 4 463 58
AK389577.1 - 2 - 3 190 58
AK399061.1 - 3 - 4 421 58
AK400531.1 - 3 - 4 461 58
DQ266356.1 - 3 - 4 456 58
EU650785.1 - 3 - 4 375 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

ethanol catabolic process
oxidation-reduction process
glyceraldehyde-3-phosphate dehydrogenase (NAD+) (non-phosphorylating) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0