This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss03388311_m1
See other PLAGL1 GE Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 21299830 - 21371070 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss03388311_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PLAGL1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001044545.1
NP_001038010.1
2 - 3
537
81
XM_005654368.3
XP_005654425.1
1083
81
XM_005654371.3
XP_005654428.1
1047
81
XM_013984759.2
XP_013840213.1
1218
81
XM_013984762.2
XP_013840216.1
1305
81
XM_013984764.2
XP_013840218.1
1005
81
XM_013984767.2
XP_013840221.1
1186
81
XM_021084039.1
XP_020939698.1
5555
81
XM_021084045.1
XP_020939704.1
5835
81
XM_021084053.1
XP_020939712.1
8141
81
XM_021084059.1
XP_020939718.1
2063
81
XM_021084064.1
XP_020939723.1
6662
81
XM_021084067.1
XP_020939726.1
1093
81
XM_021084080.1
XP_020939739.1
6213
81
XM_021084087.1
XP_020939746.1
5922
81
XM_021084092.1
XP_020939751.1
5954
81
XM_021084099.1
XP_020939758.1
5615
81
XM_021084106.1
XP_020939765.1
5370
81
XM_021084114.1
XP_020939773.1
5754
81
XM_021084121.1
XP_020939780.1
2031
81
XM_021084128.1
XP_020939787.1
5601
81
XM_021084156.1
XP_020939815.1
5491
81
XM_021084163.1
XP_020939822.1
5453
81
XM_021084171.1
XP_020939830.1
5438
81
XM_021084176.1
XP_020939835.1
971
81
XM_021084182.1
XP_020939841.1
1520
81
GenBank mRNA
DQ288899.1
-
2 - 3
537
81
FJ746562.1
-
2 - 3
537
81
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
3' Most
Recommended Assay
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of transcription from RNA polymerase II promoter
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0