This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss03381003_u1
See other TRA2B GE Assays ›
Gene Symbol:
TRA2B
Gene Name:
transformer 2 beta homolog
Gene Aliases:
SFRS10
Chromosome Location:
Chr.13: 123760008 - 123780919 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss03381003_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRA2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077221.1 NP_001070689.1 6 650 90
XM_021068630.1 XP_020924289.1 1102 90
XM_021068632.1 XP_020924291.1 1094 90
XM_021068633.1 XP_020924292.1 670 90
GenBank mRNA AK238371.1 - 6 808 90
AK239259.1 - 6 803 90
AK351209.1 - 6 802 90
AY610165.1 - 6 794 90
DQ972957.1 - 6 650 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Amplicon spans exons and probe does not span exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0